Order Entry
Germany
ContactUsLinkComponent
Cold Trap Bath, WCT-40
Cold Trap Bath, WCT-40
Catalog # WITGDH.WCT00140
Supplier:  witeg Labortechnik
New Product
Cold Trap Bath, WCT-40
Catalog # WITGDH.WCT00140
Supplier:  witeg Labortechnik
New Product

Specifications

  • Capacity:
    10 L
  • Description:
    Cold trap bath, weight: 67 kg, refrigerant: R-404A
  • Pk:
    1

Specifications

About this item

Ideal for collecting evaporation vapor or hazardous vapor from vacuum line.

  • Stainless steel bath
  • Powder-coated steel, double painted and baked, 20 mm polyurethane-insulation
  • Digital timer function (99 hr 59 min): delay time and operation time can be controlled
  • Users self-compensation function to control difference between real temperature and indicated temperature
  • Drain valve mounted
  • Backlight LCD display
  • CE certified and unique serial number for tracing

Safety mechanism:
Over-current protection
Sensor error detection and leakage breaker
Locking mode

Controller: Digital PID controller with Jog-Shuttle-switch (turn + push)

Temperature range: −65 up to −80 °C
Refrigerator: 370 W
Internal dimensions: 220×220×210 mm
External dimensions: 377×453×705 mm
Power supply: 230 V (AC), 50/60 Hz

Delivery: Includes cold trap WCT, 2 glass cold traps, power cable, instruction manual.