Order Entry
Germany
ContactUsLinkComponent
Glucokinase Activator, Cpd A ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Glucokinase Activator, Cpd A ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 346021-5MG
CAS Number:  
Glucokinase Activator, Cpd A ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 346021-5MG
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1
  • Seq:
    2210
  • Pk Info:
    5 mg

Specifications

About this item

 :
 :346.43 g/mol
Storage Temperature:Refrigerator
 :M-Clarity™ quality level = MQ100
CAS Number:603108-44-7
Formula:C₁₄H₁₄N₆OS₂