Specifications
- Pk:100
- Description:Caspase-3/7 activity flow cytometry kit, green
Specifications
About this item
For detection of active caspase-3 in mammalian cells undergoing apoptosis - 100 tests.
- TF2-DEVD-FMK detects caspase-3/7 activity
- Excitation wavelength: 503 nm
- Emission wavelength: 525 nm
- Extinction coefficient: 75,000 cm⁻¹M⁻¹
- Easily scalable for high-throughput assays
- Verified for use with flow cytometry
Efficiently detect active caspase-3 in mammalian cells undergoing apoptosis by using the Caspase-3/7 activity flow cytometry kit, green. This reagent kit uses a quick and simple staining protocol that is highly sensitive and compatible with standard flow cytometers.
Caspase-3 and caspase-7 have a proteolytic substrate selectivity for the peptide sequence Asp-Glu-Val-Asp (DEVD). This kit includes TF2-DEVD-FMK a cell-permeable and non-toxic fluorescent substrate as well as an assay buffer and propidium iodide. When added to apoptotic cells, TF2-DEVD-FMK irreversibly binds to activated caspase-3, resulting in the retention of the green fluorescent reagent within cells that can be detected by flow cytometry.
Caspases are a family of proteases that play a central role in cellular apoptosis through the cleavage of select proteins, and result in the disassembly of the cell (Thornberry & Lazebnik Science, 1998). This process is switched on by pro-apoptotic signals from death receptors or stimuli such as cytotoxic reagents, which activate the initiator caspases and lead to activation of the effector caspases, caspase-3 and caspase-7 (Thornberry & Lazebnik Science, 1998). Caspase-3/7 Activity Flow Cytometry Kit, Green is useful for screening caspase-3 inhibitors and quantifying apoptotic cells containing active caspase-3.
CAS No: 67-68-5