ContactUsLinkComponent
ErythroClear™ Magnet
ErythroClear™ Magnet
Catalog # STMC01737
undefined
ErythroClear™ Magnet
Catalog # STMC01737
Specifications
  • Description:
    ErythroClear™ Magnet
  • Pk:
    1
Specifications
About this item
Perform total nucleic acid extraction from a variety of sample types using the ErythroClear™ magnet.
  • Magnet for column-free immunomagnetic separation
  • Process up to 16 standard polypropylene microcentrifuge tubes at a time
Perform total nucleic acid extraction from a variety of sample types, including whole blood and cell suspensions (PBMCs, hPSCs, cultured cells), using the ErythroClear™ magnet in combination with EasySep™ reagents. Use the ErythroClear™ magnet to process up to 16 standard polypropylene microcentrifuge tubes at a time (8 samples per side).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').