ContactUsLinkComponent
EasyPlate™ EasySep™ Magnet
EasyPlate™ EasySep™ Magnet
Catalog # STMC18102
undefined
EasyPlate™ EasySep™ Magnet
Catalog # STMC18102
Specifications
  • Description:
    EasyPlate™ EasySep™ magnet
  • Pk:
    1
Specifications
About this item
The EasyPlate™ EasySep™ Magnet is designed for cell separation procedures using selected EasySep™ reagents when processing a small number of cells (up to 2×10⁷ cells per well).
  • Immunomagnetic column-free magnet
  • Designed to hold a round-bottom, non-tissue culture treated 96-well plate
The EasyPlate™ EasySep™ magnet generates a high-gradient magnetic field that is strong enough to separate cells labeled with EasySep™ D magnetic particles, without the use of columns. The EasyPlate™ EasySep™ magnet is designed to hold a round-bottom, non-tissue culture treated 96-well plate.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').