OptiPrep™ Density Medium
Catalog # STMC07820
Supplier: STEMCELL Technologies
Specifications
- Description:OptiPrep™ density medium (in bottle)
- Pk:250 mL
Specifications
About this item
Density gradient medium for the isolation of viruses, organelles, macromolecules, or cells.
- Use to form continuous or discontinuous gradients
- Customise separation characteristics for isolation of viruses, organelles, macromolecules, or cells
- Non-ionic, non-toxic to cells, isoosmotic, and metabolically inert
Simplify the isolation and purification of macromolecules, viruses, a wide range of cell types, and organelles such as nuclei, mitochondria, endosomes, or exosomes by using OptiPrep™ medium. This flexible, versatile, and gentle density gradient medium is non-ionic, iodixanol-based (60% w/v) and has a density of 1,320 ±0,001 g/ml. OptiPrep™ has been used clinically in X-ray contrast imaging and has been shown to have lower toxicity when compared to ionic gradient compounds such as metrizamide.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').