Circular Vinyl Labels for BradyJet™ J7300 Inkjet Label Printer
Supplier: Brady
New Product
Certificates
About this item
General-purpose vinyl labels feature a specially formulated topcoat for improved durability and colour. For use on smooth and textured and rough surfaces.
- Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
- Works well in low temperatures with a service temperature of −40 to 60 °C
- Inkjet receptive vinyl material
- Designed for indoor/outdoor use
- Abrasion-resistant, oil-resistant, outdoor durability, solvent/chemical resistant
- Matt finish
Low-shrink vinyl with ultra-aggressive adhesive is ideal for circular machine gauge labels common to Lean/5S applications. Enhanced topcoat works with durable J7300 inks to protect printed text and graphics from smudging and harsh conditions. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 colour label inkjet printer. Ink cartridges are required to print on this material; use part numbers P-CMY-1 and P-K-1.