Order Entry
Germany
ContactUsLinkComponent
Trypan blue 0.4% in PBS
Health
Trypan blue 0.4% in PBS
Catalog # STMC07050
CAS Number:  
undefined
Health
Trypan blue 0.4% in PBS
Catalog # STMC07050
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :960.82 g/mol
 :GHS08
Storage Temperature:Ambient
CAS Number:72-57-1
Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
MDL Number:MFCD00003969