Order Entry
Germany
ContactUsLinkComponent
 
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)]
Catalog # STMC75003
CAS Number:  
undefined
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)]
Catalog # STMC75003
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :557.467 g/mol
CAS Number:150347-59-4
 :MFCD31812843
Formula:C₂₉H₁₉NO₁₁
MDL Number:MFCD00037466