Showa DURACoil 346 Cut Resistant Gloves
: Showa
Don't compromise on flexibility when seeking highly durable cut-resistant gloves.
- Cut-resistant and abrasion-resistant
- Form-fitting
- Reusable
- Machine washable
- Flexible and breathable
- Increased dexterity
- Seamless knit
The DURACoil 346 utilises an advanced HPPE reinforced lining to provide cut and abrasion resistance, without sacrificing dexterity. The natural rubber latex palm dip is liquid-proof for added comfort and precision. Even with tasks on a smaller scale, such as component assembly, the 346 allows for full range of movement and control.
: Cat II. EN 388:2016+A1:2018, 2X42C.
: Gloves are packaged 10 pair per polybag, 12 polybags per case (120 total pair per case).
- Catalog No:
- BEST346-9/XL
- BEST346-8/L
- BEST346-6/S
- BEST346-7/M
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex
- Latex
- Latex
- Latex
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- HPPE
- HPPE
- HPPE
- HPPE
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile