Order Entry
Germany
ContactUsLinkComponent
Human Serum AB (Off The Clot), Gamma Irradiated
Human Serum AB (Off The Clot), Gamma Irradiated
Catalog # GEMB100-318G-100
New Product
undefined
Human Serum AB (Off The Clot), Gamma Irradiated
Catalog # GEMB100-318G-100
New Product

Specifications

  • Serum format:
    Frozen
  • Sterility:
    Sterile filtered
  • Description:
    Human serum AB (off the clot) - gamma irradiated
  • Shipping conditions:
    Ships on dry ice
  • Origin:
    US
  • Storage temperature:
    Store frozen at −20...−10 °C
  • Pack type:
    Bottle
  • Pk:
    100 mL

Specifications

About this item

Off the clot human serum AB is collected from normal healthy male donors at FDA-licensed facilities in the United States. As a natural off the clot product, this material contains no anticoagulants/preservatives. All donor units are tested for viral markers and found to be non-reactive.

  • Supports clinical and commercial applications
  • Aseptic assurance system
  • Gamma irradiated