Anti-4 HNE Rabbit Polyclonal Antibody (AbBy Fluor® 647)
BOSSBS-6313R-BF647
: Bioss
New Product
- Antibody type:Primary
- Antigen name:4 Hydroxynonenal
- Antigen symbol:4 HNE
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Monkey,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:4-Hydroxynonenal|4-Hydroxy-2-Nonenal|4-HNE|4Hydroxynonenal|4HNE|E-4-Hydroxynonenal-dimethylacetal|4 HNE|4-HNE-DMA
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,02% Proclin300 and 50% glycerol
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by Protein A
- Pk:100 µl
Aldehydic products of lipid peroxidation, such as 4 hydroxynonenal (4 HNE), have been implicated in the etiology of pathological changes under oxidative stress as a key mediator of oxidative stress induced cell death. It is a stable product of lipid peroxidation, is proarrhythmic and may contribute to the cytotoxic effects of oxidative stress 4-HNE has been hypothesised to play a key role in cell signal transduction, in a variety of pathways from cell cycle events to cellular adhesion.
- Application dilution: WB - 1:300 to 5000; IF (IHC-P) - 1:50 to 200
Type: Primary
Antigen: 4 HNE
Clonality: Polyclonal
Clone:
Conjugation: AbBy Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, rat, monkey