Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Earplugs
Disposable Earplugs
Accessories for Disposable Earplug, Bilsom 303
Accessories for Disposable Earplug, Bilsom 303
Accessories for Disposable Earplug, Bilsom 303
Catalog # DALL1006203-AM
Supplier:
HONEYWELL SAFETY
New Product
Expand image
Accessories for Disposable Earplug, Bilsom 303
Catalog # DALL1006203-AM
Supplier:
HONEYWELL SAFETY
New Product
Specifications
Colour:
Black
Description:
Ear plug dispenser for use with earplugs
Pk:
1
Specifications
About this item
Product Details
Product Details