Order Entry
Germany
ContactUsLinkComponent
Sodium Cacodylate, 0,1 M Buffer Solution, pH 6,8
Sodium Cacodylate, 0,1 M Buffer Solution, pH 6,8
  J62202.AE
 :  
Sodium Cacodylate, 0,1 M Buffer Solution, pH 6,8
  J62202.AE
 :  

 

  • Pk:
    100 mL
  • CAS No.:
    6131-99-3
  • Description:
    Sodium cacodylate, 0,1 M buffer solution, pH 6,8
  • MDL No.:
    MFCD00149079
  • Storage temperature:
    2...8 °C
  • Physical form:
    Liquid

 

 

Liquid.

CAS No.: 6131-99-3
Storage Temperature: 2 to 8 ℃
Physical Form: Liquid
MDL No.: MFCD00149079