ContactUsLinkComponent
Graduated Receiving Cylinder
Graduated Receiving Cylinder
Catalog # PAAR195234
Supplier:  Anton Paar
New Product
Graduated Receiving Cylinder
Catalog # PAAR195234
Supplier:  Anton Paar
New Product
Specifications
  • Capacity:
    100 ml
  • Description:
    Receiving cylinder with Viton O-ring, magnetic base
  • Pk:
    1
Specifications
About this item
Receiving cylinder with Viton O-ring,100 ml. Compatible with Diana 700, Diana 300.
Caution: Not suitable for solvents that dissolve rubber seals, e.g. acetone.