Supelco® TraceCERT® Organic Reference Standard, Benzo[a]pyrene solution
Catalog # CRM48665
Supplier: Merck
CAS Number:
Specifications
- Concentration:200 μg/ml in methylene chloride
- Pk:1 mL
- Standards:Certified reference material, TraceCERT®
- CAS No.:50-32-8
- Description:Benzo[a]pyrene solution
- Storage conditions:2...30 °C
- Hazard class description:H315 - H319 - H336 - H350
- Shipping conditions:STD
- MDL No.:MFCD00003602
- M-Clarity quality level:MQ100
- Molecular weight:252,31
Specifications
About this item
- GHS signal: GHS08
Caution: P202 - P261 - P264 - P302 + P352 - P305 + P351 + P338 - P308 + P313