mFluor™ Violet 500-Wheat Germ Agglutinin (WGA) Conjugate
Catalog # AATB25452
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- Pk:1 mg
- Description:mFluor™ Violet 500-Wheat Germ Agglutinin (WGA) conjugate
Specifications
About this item
mFluor™ Violet 500 labelled WGA is well-excited by the violet laser, emitting a bright green fluorescence at 501 nm.
- Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Caution: For research use only.