ContactUsLinkComponent
Gelite™ 50 bp DNA Ladder
Gelite™ 50 bp DNA Ladder
Catalog # AATB17750
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
undefined
Gelite™ 50 bp DNA Ladder
Catalog # AATB17750
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    0,5 mL
  • Description:
    Gelite™ 50 bp DNA ladder
Specifications
About this item
The Gelite™ 50 bp DNA Ladder is ideal for sizing and approximating the mass of double-stranded DNA within the 50 to 1500 bp range on agarose or polyacrylamide gels.
  • Stable for 12 months
Quality tested in-house.
Caution: For research use only.