Gelite™ 100 bp DNA Ladder
Catalog # AATB17753
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Pk:0,5 mL
- Description:Gelite™ 100 bp DNA ladder
Specifications
About this item
The Gelite™ 100 bp DNA Ladder is designed for sizing and estimating the mass of double-stranded DNA and PCR products within the 100 to 1500 bp range on agarose or polyacrylamide gels.
- Stable for 12 months
Quality tested in-house.
Caution: For research use only.