ContactUsLinkComponent
Amplite® Fluorimetric Neutrophil Elastase Activity Assay Kit
Amplite® Fluorimetric Neutrophil Elastase Activity Assay Kit
Catalog # AATB11327
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Amplite® Fluorimetric Neutrophil Elastase Activity Assay Kit
Catalog # AATB11327
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    100 Tests
Specifications
About this item
Designed for consistent and dependable results. The Amplite® fluorimetric neutrophil elastase activity assay kit provides a robust and accurate method for quantifying neutrophil elastase (NE) activity.
  • Ex/Em= 360/470 nm
  • Consistent results
  • Six-month shelf life
The kit’s straightforward protocol is optimised for the sensitive detection of NE in blood and plasma samples and is suitable for screening and characterising chemotactic agents triggering neutrophil stimulation.
Delivery: Shipped with blue ice.
Caution: Research use only.