Supelco® PESTANAL® Organic Reference Standard, 2,4,5-Trichlorophenol
Catalog # 36513-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:95-95-4
- Description:2,4,5-Trichlorophenol
- Storage conditions:RT
- Hazard class description:H302 - H315 - H319 - H410
- Shipping conditions:STD
- MDL No.:MFCD00002170
- Melting point:67...69 °C (lit.)
- Formula:Cl₃C₆H₂OH
- Molecular weight:197,45
- Flash point:133,0 °C - closed cup
- Boiling point:248 °C/740 mm Hg (lit.)
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P273 - P301 + P312 + P330 - P302 + P352 - P305 + P351 + P338