Order Entry
Germany
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, 4,4′-Methylenebis(N,N-Di Methylaniline)
Supelco® TraceCERT® Organic Reference Standard, 4,4′-Methylenebis(N,N-Di Methylaniline)
Catalog # 07559-50MG
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, 4,4′-Methylenebis(N,N-Di Methylaniline)
Catalog # 07559-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • CAS No.:
    101-61-1
  • Description:
    4,4′-Methylenebis(N,N-dimethylaniline)
  • Storage conditions:
    15...25 °C
  • Hazard class description:
    H350 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00008317
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    N,N,N′,N′-Tetramethyl-4,4′-diaminodiphenylmethane; N,N,N′,N′-Tetramethyl-4,4′-methylenedianiline; Arnolds base; Michlers base
  • Melting point:
    88...89 °C (lit.)
  • Formula:
    CH₂[C₆H₄N(CH₃)₂]₂
  • Molecular weight:
    254,37
  • Flash point:
    178 °C - closed cup

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P201 - P202 - P273 - P280 - P308 + P313 - P391