HIS Lite™ iFluor® 568 Tris NTA Chelator
Catalog # AATB12657
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Pk:100 µG
- Description:HIS Lite™ iFluor™ 568 tris NTA chelator
Specifications
About this item
iFluor® 568 Tris-NTA Chelator is used as a sensitive fluorescent probe for detecting polyhistidine-labelled proteins in combination with the addition of a heavy metal ion (such as Ni²⁺) in cells, solution and solid surfaces.
- Stable for 24 months
Quality tested in-house.
Caution: For research use only.