About this item
This clear liner-less cryogenic tape for already frozen surfaces is provided with innovative GatorCUT technology, making it effortlessly hand-cuttable. Perfect for identifying frozen samples and containers without concealing their contents, ensuring they remain visible.
- Temperature range: –196 to +121 °C
- Applicable on frozen surfaces, as low as –80 °C
- Ideal for storage in ultra-low temperature conditions
- Liner-free with GatorCUT technology; portable without the need for cutting instruments
- Transparent design ensures contents of labeled containers remain visible
This tape is designed to stick to frozen surfaces, including cryo vials, boxes, and long-term storage in liquid nitrogen or deep-freeze laboratory freezers without losing the adhesive properties. The cryogenic tape also resists high temperatures, making it a versatile option for professionals in diverse environments. Ideal for use in scientific applications where materials need to be stored at very low or high temperatures from –196 to +120 °C.
The tape also features GatorCUT technology, which allows for precise and accurate cutting of the tape to the desired length. This makes it easy to label and identify samples or storage containers in cryogenic environments. Offered in a clear format, this self-wound tape allows for clear sample visualisation.