Order Entry
Germany
ContactUsLinkComponent
Supelco® Organic Reference Standard, F.A.M.E. Mix C8 - C24
Supelco® Organic Reference Standard, F.A.M.E. Mix C8 - C24
Catalog # CRM18918
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, F.A.M.E. Mix C8 - C24
Catalog # CRM18918
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    F.A.M.E. mix C8 - C24
  • Storage conditions:
    –10...–25 °C
  • Hazard class description:
    H410
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ100
  • Pk Info:
    Ampule of 100 mg

Specifications

About this item

This certified reference material is useful for determining relative retention times and approximating response factors.

  • GHS signal: GHS07
Caution: P273 - P391 - P501