Order Entry
Germany
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, Benfuracarb
Supelco® TraceCERT® Organic Reference Standard, Benfuracarb
Catalog # CRM06035-100MG
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, Benfuracarb
Catalog # CRM06035-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    82560-54-1
  • Description:
    Benfuracarb
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H301 + H331 - H361f - H410
  • MDL No.:
    MFCD00210247
  • M-Clarity quality level:
    MQ300
  • Molecular weight:
    410,53

Specifications

About this item

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. Benfuracarb certified reference material (CRM) may also be used as following:

  • Residue analysis of 18 insecticides in water and sediment samples collected from aquatic areas around the rice paddy fields by chromatography tandem mass spectrometry
  • Analysis of 168 pesticides in paprika using the modified dry ice-partitioning QuEChERS method combined with LC-MS/MS
  • Simultaneous determination of 18 carbamate pesticide residues from whole chestnut and pine nut samples using ultra-performance liquid chromatography coupled with tandem mass spectrometry (UPLC-MS-MS) following a gel permeation chromatography (GPC) clean-up
  • Development and validation of a multi-residue analysis method for detecting 70 pesticides belonging to different classes from drinking water samples using a polymeric sorbent for solid phase extraction (SPE) and gas and liquid chromatography coupled to tandem mass spectrometry (GC-MS/MS and LC-MS/MS) for quantification
  • Detection of 15 organic nitrogen-containing pesticides from vegetable samples using matrix solid-phase dispersion (MSPD) along with gas chromatography-Tandem mass spectrometry (GC-MS-MS)
  • Simultaneous analysis of 27 organophosphorus, organochlorine, pyrethroid, and carbamate pesticides in Radix astragali by microwave-assisted or dispersive-solid phase extraction in combination with GC-MS
Caution: P202 - P261 - P264 - P273 - P301 + P310 - P304 + P340 + P311