Order Entry
Germany
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, Tebufenozide
Supelco® TraceCERT® Organic Reference Standard, Tebufenozide
Catalog # CRM01373-50MG
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, Tebufenozide
Catalog # CRM01373-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    112410-23-8
  • Description:
    Tebufenozide
  • Storage conditions:
    15...25 °C
  • Hazard class description:
    H411
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00467963
  • M-Clarity quality level:
    MQ300
  • Molecular weight:
    352,47

Specifications

About this item

It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Tebufenozide certified reference material (CRM) may also find the following uses:

  • Analysis of trifloxystrobin tebufenozide and halofenozide by Micellar electrokinetic capillary chromatography (MEKC) in tomato celery and apple juice samples
  • Measurement of the residue levels of tebufenozide in cabbage and soil by reversed-phase high-performance liquid chromatography (HPLC)
  • Determination of four diacylhydrazine insecticide residues in apple grape cucumber tomato and spinach samples by QuEChERS method using ultra-performance liquid chromatography coupled with tandem mass spectrometry
  • Development and validation of a multi-residual analytical method based on SPE followed by LC-MS for the simultaneous estimation of 14 pesticides in surface and groundwaters
  • Multi-residue analysis of 200 pesticides and pesticide metabolites in honeybees by gas and liquid chromatography coupled with tandem mass spectrometry
Certifications: It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Tebufenozide certified reference material (CRM) may also find the following uses:
  • Analysis of trifloxystrobin tebufenozide and halofenozide by Micellar electrokinetic capillary chromatography (MEKC) in tomato celery and apple juice samples
  • Measurement of the residue levels of tebufenozide in cabbage and soil by reversed-phase high-performance liquid chromatography (HPLC)
  • Determination of four diacylhydrazine insecticide residues in apple grape cucumber tomato and spinach samples by QuEChERS method using ultra-performance liquid chromatography coupled with tandem mass spectrometry
  • Development and validation of a multi-residual analytical method based on SPE followed by LC-MS for the simultaneous estimation of 14 pesticides in surface and groundwaters
  • Multi-residue analysis of 200 pesticides and pesticide metabolites in honeybees by gas and liquid chromatography coupled with tandem mass spectrometry
Caution: P273 - P391 - P501