Supelco® TraceCERT® Organic Reference Standard, Metal mix I for cannabis testing
91539-100ML
: Merck
:
- Concentration:12 elements, (in 8% nitric acid and 0,05% HF)
- Pk:100 mL
- Standards:TraceCERT®
- Description:Metal mix I for cannabis testing
- Storage conditions:No temperature limit
- Hazard class description:H290 - H314 - H317 - H350 - H411
- Shipping conditions:STD
- M-Clarity quality level:MQ300
: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
: P202 - P273 - P280 - P303 + P361 + P353 - P304 + P340 + P310 - P305 + P351 + P338