Supelco® Organic Reference Standard, Trifluralin
Catalog # 90662-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- CAS No.:1582-09-8
- Description:Trifluralin
- Storage conditions:15...25 °C
- Hazard class description:H302 - H317 - H351 - H410
- MDL No.:MFCD00055241
- M-Clarity quality level:MQ100
- Synonym(s):2,6-Dinitro-N,N-dipropyl-4-trifluoromethylaniline
- Molecular weight:335,28
Specifications
About this item
Caution: P202 - P273 - P280 - P301 + P312 - P302 + P352 - P308 + P313