Supelco® TraceCERT® Organic Reference Standard, Imazamethabenz-methyl
Imazamethabenz-methyl, Certified reference material, TraceCERT®, 50 mg
Catalog # 90556-50MG
Supplier: Merck
Supelco® TraceCERT® Organic Reference Standard, Imazamethabenz-methyl
Catalog # 90556-50MG
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The imazamethabenz-methyl CRM may also find following uses:
- Study the photolysis of imazamox imazapic imazaquin and imazamethabenz-methyl as functions of wavelength pH and natural organic matter in an aqueous solution
- Comparative study of the extraction of imazamethabenz-methyl from agricultural soils followed by its chromatographic determination using HPLC
- Investigate the absorption and translocation of fenoxaprop-ethyl and imazamethabenz-methyl in wild oat plants grown under different conditions of temperature and light intensity using ¹⁴C tracer techniques
- Simultaneous determination of p- and m-imazamethabenz-methyl enantiomers using a protein chiral selector based on a direct chiral HPLC method
- To develop a multiresidue method for the determination of post-emergence herbicides in water samples by SPE coupled with HPLC/ESI/MS in positive ionisation mode
Specifications
Specifications
- Pk:50 mg
- Standards:Certified reference material, TraceCERT®
- Description:Imazamethabenz-methyl
- Cat. No.:90556-50MG
- M-Clarity quality level:MQ300
- Storage conditions:15...25 °C
- MDL No.:MFCD00144469
- Molecular weight:288,34
- CAS No.:81405-85-8
- Shipping conditions:STD
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



