Order Entry
Germany
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, α-Endosulfan
Supelco® TraceCERT® Organic Reference Standard, α-Endosulfan
Artikelnr. 74119-50MG
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, α-Endosulfan
Artikelnr. 74119-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    959-98-8
  • Description:
    α-Endosulfan
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H300 + H330 - H312 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00151174
  • M-Clarity quality level:
    MQ300
  • Molecular weight:
    406,93

Specifications

About this item

  • GHS signal: GHS06, GHS09
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P260 - P264 - P273 - P280 - P302 + P352 + P312 - P304 + P340 + P310