Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
MTS reagent
MTS reagent 5 mM in aqueous solution, ReadiUse™
MTS reagent 5 mM in aqueous solution, ReadiUse™
MTS reagent 5 mM in aqueous solution, ReadiUse™
Supplier:
AAT BIOQUEST
New Product
Certificates
Bild vergrößern
MTS reagent 5 mM in aqueous solution, ReadiUse™
Supplier:
AAT BIOQUEST
New Product
About this item
CAS Number:
138169-43-4
:
Formula:
C₂₀H₁₇N₅O₆S₂
:
487.51 g/mol