Order Entry
Germany
Orders LinkContactUsLinkComponent
mFluor™ Blue 585 SE

mFluor™ Blue 585 SE

1 mg
Catalog # AATB1159
Supplier:   AAT Bioquest
mFluor™ Blue 585 SE
Catalog # AATB1159
Supplier:  AAT Bioquest
CAS Number:  
mFluor™ Blue 585 SE

1 mg

CAS Number:  
Price
469,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

mFluor™ Blue 585 dyes are an excellent alternative to RPE since they have the spectral properties equivalent to those of RPE conjugates.

  • Stable for 12 months

Quality tested in-house

Caution: For Research Use Only

  • Molecular Weight:835.92 g/mol
Specifications

Specifications

  • Pk:
    1 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.