0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
DBCO-PEG5-NHS ester
DBCO-PEG5-NHS ester
DBCO-PEG5-NHS ester
Catalog # AATB4526
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
DBCO-PEG5-NHS ester
Catalog # AATB4526
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
Product Details