Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Cy5 aldehyde

1 mg
Catalog # AATB9012
Supplier:   AAT Bioquest
Cy5 aldehyde
Catalog # AATB9012
Supplier:  AAT Bioquest
CAS Number:  
Cy5 aldehyde

1 mg

CAS Number:  
Price
288,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Cy5 aldehyde is a reactive red fluorescent dye that can react with an amine, hydrazine or hydroxylamine.

  • Stable for 12 months

Quality tested in-house

 : For Research Use Only

  • Molecular Weight:945.04 g/mol
Specifications

Specifications

  • Pk:
    1 mg

Specifications