Order Entry
Germany
Orders LinkContactUsLinkComponent
Ac-DEVD-aminoluciferin

Ac-DEVD-aminoluciferin

1 mg
Catalog # AATB13210
Supplier:   AAT Bioquest
Ac-DEVD-aminoluciferin
Catalog # AATB13210
Supplier:  AAT Bioquest
CAS Number:  
Ac-DEVD-aminoluciferin

1 mg

CAS Number:  
Price
134,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Ac-DEVD-aminoluciferin is a luminogenic substrate that measures caspase-3 and -7 activities in apoptotic cells.

  • Stable for 24 months

Quality tested in-house

Caution: For Research Use Only

  • Formula:C₃₁H₃₇N₇O₁₃S₂
  • Molecular Weight:779.79 g/mol
Specifications

Specifications

  • Pk:
    1 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.