ContactUsLinkComponent
Beta Sterile Kits
New Product
undefined
Beta Sterile Kits
New Product
  • Catalog No:
  • ISOF80451XXXL
  • ISOF80451XL
  • ISOF80451XXL
  • ISOF80451M
  • ISOF80451S
  • ISOF80451L
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Anti-static:
  • +
  • +
  • +
  • +
  • +
  • +
  • Cleanroom class:
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4