Anti-TRP1 Mouse Monoclonal Antibody [clone: TYRP1/3284]
Catalog # ANTIA253431-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Glycoprotein 75
- Antigen symbol:TRP1
- Clonality:Monoclonal
- Clone:TYRP1/3284
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P17643
- Antigen synonyms:CAS2|Catalase B|6-dihydroxyindole-2-carboxylic acid oxidase|GP75|CATB|b-PROTEIN|5,6 dihydroxyindole 2 carboxylic acid oxidase|DHICA oxidase|5
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:75 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, within amino acids 257 - 377, of human TYRP1 protein. The exact sequence is proprietary
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [TYRP1/3284] antibody to TRP1 for ELISA and IHC-P with samples derived from human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: TRP1
Clonality: Monoclonal
Clone: TYRP1/3284
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human