Order Entry
Germany
ContactUsLinkComponent
Anti-Transthyretin Mouse Monoclonal Antibody [clone: CPTC-TTR-1]
Anti-Transthyretin Mouse Monoclonal Antibody [clone: CPTC-TTR-1]
Catalog # ANTIA253403-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Transthyretin Mouse Monoclonal Antibody [clone: CPTC-TTR-1]
Catalog # ANTIA253403-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Amyloid polyneuropathy
  • Antigen symbol:
    Transthyretin
  • Clonality:
    Monoclonal
  • Clone:
    CPTC-TTR-1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P02766
  • Antigen synonyms:
    Amyloidosis I|HEL111|CTS1|Dystransthyretinemic hyperthyroxinemia|Dysprealbuminemic euthyroidal hyperthyroxinemia|CTS|Carpal tunnel syndrome 1|ATTR|Epididymis luminal protein 111
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    15 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human Transthyretin protein
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [CPTC-TTR-1] antibody to Transthyretin for WB with samples derived from human.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1 to 2 µg/ml

Type: Primary
Antigen: Transthyretin
Clonality: Monoclonal
Clone: CPTC-TTR-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human