Anti-TRP1 Mouse Monoclonal Antibody [clone: TA99 + TYRP1/807]
ANTIA253436-100
New Product
- Antibody type:Primary
- Antigen name:5,6 dihydroxyindole 2 carboxylic acid oxidase
- Antigen symbol:TRP1
- Clonality:Monoclonal
- Clone:TA99 + TYRP1/807
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:Multiple
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P17643
- Antigen synonyms:CAS2|Catalase B|6-dihydroxyindole-2-carboxylic acid oxidase|GP75|CATB|Glycoprotein 75|b-PROTEIN|DHICA oxidase
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:75 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Clone TA99: SK-MEL-23 cells. Clone TYRP1/807: Recombinant full-length human TYRP1 protein.
- Tested applications:IHC
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [TA99 + TYRP1/807] antibody to TRP1 for Flow Cytometry, IF, WB and IHC with samples derived from human and mouse.
- Validated Applications: Flow Cytometry, IF, WB, IHC
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: TRP1
Clonality: Monoclonal
Clone: TA99 + TYRP1/807
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human, Mouse