Order Entry
Germany
ContactUsLinkComponent
Anti-NMDAR2A Rabbit Monoclonal Antibody [clone: ARC0410]
Anti-NMDAR2A Rabbit Monoclonal Antibody [clone: ARC0410]
Catalog # ANTIA308430-100
Supplier:  Antibodies.com
New Product
undefined
Anti-NMDAR2A Rabbit Monoclonal Antibody [clone: ARC0410]
Catalog # ANTIA308430-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    EPND
  • Antigen symbol:
    NMDAR2A
  • Clonality:
    Monoclonal
  • Clone:
    ARC0410
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q12879
  • Antigen synonyms:
    hNR2A|LKS|GRIN2A|GluN2A|GRIN 2A|Glutamate [NMDA] receptor subunit epsilon-1|FESD|Glutamate receptor|Glutamate receptor ionotropic N methyl D aspartate 2A
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    180 kDa
  • Sequence:
    RELDLSRPSRSISLKDRERLLEGNFYGSLFSVPSSKLSGKKSSLFPQGLEDSKRSKSLLPDHTSDNPFLHSHRDDQRLVIGRCPSDPYKHSLPSQAVNDSY
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 1300-1400 of human NMDAR2A (Q12879)
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit monoclonal [ARC0410] antibody to NMDAR2A for WB and IHC with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC
  • Recommended dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200

Type: Primary
Antigen: NMDAR2A
Clonality: Monoclonal
Clone: ARC0410
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat