Order Entry
Germany
ContactUsLinkComponent
 
Anti-Superoxide Dismutase 4/CCS Rabbit Polyclonal Antibody
Artikelnr. ANTIA9998-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Superoxide Dismutase 4/CCS Rabbit Polyclonal Antibody
Artikelnr. ANTIA9998-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Superoxide Dismutase 4/CCS
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# O14618
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
  • Molecular weight:
    37 kDa
  • Sequence:
    MASDSGNQGTLCTLEFAVQMTCQSCVDAVRKSLQGVAGVQDVEVHLEDQMVLVHTTLPSQEVQALLEGTGRQAVLKGMGSGQLQNLGAAVAILGGPGTVQGVVRFLQLTPERCLIEGTIDGLEPGLHGLHVHQYGDLTNNCNSCGNHFNPDGASHGGPQDSDRHRGDLGNVRADADGRAIFRMEDEQLKVWDVIGRSLIIDEGEDDLGRGGHPLSKITGNSGERLACGIIARSAGLFQNPKQICSCDGLTIWEERGRPIAGKGRKESAQPPAHL
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-274 of human CCS (NP_005116.1)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to Superoxide Dismutase 4/CCS for WB with samples derived from human.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:200

Type: Primary
Antigen: Superoxide Dismutase 4/CCS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human