Anti-NMDAR2C Rabbit Polyclonal Antibody
Catalog # ANTIA98698-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:GluN2C
- Antigen symbol:NMDAR2C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q14957
- Antigen synonyms:Glutamate Receptor Ionotropic N Methyl D Aspartate 2C|N Methyl D Aspartate Receptor Subtype 2C|N Methly D Aspartate Receptor Channel Subunit Epsilon 3|Grin2c|N-methyl D-aspartate receptor subtype 2C|N methyl D aspartate receptor subunit 2C|Glutamate receptor, ionotropic, N methyl D aspartate 2C|Glutamate receptor ionotropic, NMDA 2C|Glutamate [NMDA] receptor subunit epsilon-3
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:134 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human NMDAepsilon3 (amino acids 937-986)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to NMDAepsilon3 for IHC and ELISA with samples derived from human.
- Validated applications: IHC, ELISA
- Recommended dilutions: IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: NMDAR2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human