Order Entry
Germany
ContactUsLinkComponent
 
Anti-CDK2 (phospho Tyr15) Rabbit Antibody [clone: ARC1550]
  ANTIA306143-100
New Product
undefined
Anti-CDK2 (phospho Tyr15) Rabbit Antibody [clone: ARC1550]
  ANTIA306143-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Cdc2 related protein kinase
  • Antigen symbol:
    CDK2
  • Clonality:
    Monoclonal
  • Clone:
    ARC1550
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P24941
  • Antigen synonyms:
    CDKN2|CDK1|CDK2_HUMAN|cdc2-related protein kinase|Cdk 2|CDC28|Cell devision kinase 2|CDC2A|Cdk2
  • Modification:
    phospho Tyr15
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    34 kDa
  • Sequence:
    GTYGV
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic phosphorylated peptide around Y15 of human CDK2 (P24941).
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit monoclonal [ARC1550] antibody to CDK2 (phospho Tyr15) for WB with samples derived from Human and Mouse.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: CDK2
Clonality: Monoclonal
Clone: ARC1550
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse