Anti-DOCK1 Goat Polyclonal Antibody
Catalog # ANTIA83785-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:180 kDa protein downstream of CRK
- Antigen symbol:DOCK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:DOCK180|Dedicator of Cytokinesis 1|DOCK 1|DOCK1|Ced5|Dedicator of cytokinesis protein 1|DOCK 180|DOCK1_HUMAN
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:C-TTRKQTSVDSGIVQ
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human DOCK1 (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to DOCK1 for WB with samples derived from human.
- Validated applications: W
Type: Primary
Antigen: DOCK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human