Anti-TRP1 Rabbit Monoclonal Antibody [clone: TYRP1/8313R]
ANTIA317143-100
New Product
- Antibody type:Primary
- Antigen name:TRP1
- Antigen symbol:TRP1
- Clonality:Monoclonal
- Clone:TYRP1/8313R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P17643
- Antigen synonyms:CAS2|Catalase B|6-dihydroxyindole-2-carboxylic acid oxidase|GP75|CATB|Glycoprotein 75|b-PROTEIN|5,6 dihydroxyindole 2 carboxylic acid oxidase|DHICA oxidase|5
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without Sodium azide and carrier free
- Molecular weight:75 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 200-400) of human TYRP1 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Rabbit monoclonal [TYRP1/8313R] antibody to TRP1 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: TRP1
Clonality: Monoclonal
Clone: TYRP1/8313R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human