Anti-GRIK1 Goat Polyclonal Antibody
ANTIA83293-100
New Product
- Antibody type:Primary
- Antigen name:EAA3
- Antigen symbol:GRIK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:GluR5|GLR5|Glutamate receptor 5|GluR6|EEA3|GluR-5|Excitatory amino acid receptor 3|GluR7|Glutamate receptor
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:QCKQTHPTNSTS
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human GRIK1 (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Goat polyclonal antibody to GRIK1 for ELISA, WB and IHC with samples derived from human.
- Validated applications: ELISA, WB, IHC
Type: Primary
Antigen: GRIK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human