PlateLoc Microplate Heat Seals
Supplier: AGILENT
New Product
Certificates
About this item
Certified for use with the PlateLoc Thermal Microplate Sealer.
- Choice of aluminum-based or optically clear materials with peelable, pierceable or permanent sealants
- Seals to match most microplate materials and applications, including polypropylene, polystyrene and COC
Agilent offers a range of PlateLoc microplate heat seals made from aluminum-based or optically clear materials with peelable, pierceable or permanent sealants matching most microplate materials and applications.
Ideal for life science applications that require automated microplate heat sealing, Agilent heat seals keep the PlateLoc Thermal Microplate Sealer up and running reliably to securely heat seal your microplates.