Cyanine-5- dUTP [Cy5-dUTP] *1 mM in TE Buffer (pH 7,5)*
Supplier: AAT BIOQUEST
New Product
Certificates
About this item
This Cy5-dUTP conjugate can be used as a red fluorescence colour with SpectrumFRed™ filter set (SpectrumFRed™ is the trademark of Vysis).
- Functionally equivalent to Cyanine 5-dUTP (Perkin Elmer’s NEL579001EA)
The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labelled DNA via conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labelling, or PCR.
Caution: For research use only.