Order Entry
Germany
ContactUsLinkComponent
Anti-TRP1 Mouse Monoclonal Antibody [clone: TYRP1/1986]
Anti-TRP1 Mouse Monoclonal Antibody [clone: TYRP1/1986]
  ANTIA250257-100
New Product
undefined
Anti-TRP1 Mouse Monoclonal Antibody [clone: TYRP1/1986]
  ANTIA250257-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    GP75
  • Antigen symbol:
    TRP1
  • Clonality:
    Monoclonal
  • Clone:
    TYRP1/1986
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P17643
  • Antigen synonyms:
    CAS2|Catalase B|6-dihydroxyindole-2-carboxylic acid oxidase|CATB|Glycoprotein 75|b-PROTEIN|5,6 dihydroxyindole 2 carboxylic acid oxidase|DHICA oxidase|5
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    75 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, within amino acids 257 - 377, of human TYRP1 protein. The exact sequence is proprietary
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [TYRP1/1986] antibody to TRP1 for ELISA and IHC-P with samples derived from Human.

  • Validated applications: ELISA, IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: TRP1
Clonality: Monoclonal
Clone: TYRP1/1986
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human